1-(3,5-Dichlorophenyl)-1,2-propandione 95+%

Lieferant: Apollo Scientific

OR1885-1G OR1885-5G OR1885-25G
APOSOR1885-1GEA 0 EUR
APOSOR1885-1G APOSOR1885-5G APOSOR1885-25G
1-(3,5-Dichlorophenyl)-1,2-propandione 95+%
1-(3,5-Dichlorophenyl)-1,2-propandione
Formel: C₉H₆Cl₂O₂
Schmelzpunkt: 45…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01941316
CAS-Nummer: 386715-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR