Methyl 6-chloropyrimidine-4-carboxylate

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-(methoxycarbonyl)-1,3-diazine, 4-Chloro-6-(methoxycarbonyl)pyrimidine

OR18854-5G
APOSOR18854-5GEA 94.4 EUR
APOSOR18854-5G
Methyl 6-chloropyrimidine-4-carboxylate
Methyl 6-chloropyrimidine-4-carboxylate
Formel: C₆H₅ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08689467
CAS-Nummer: 6627-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR