N-(6-Bromhexyl)phthalimid
Lieferant: Apollo Scientific
|
Achtung
|
OR1895-1G
APOSOR1895-1GEA
21.9
EUR
APOSOR1895-1G
N-(6-Bromhexyl)phthalimid
N-(6-Bromhexyl)phthalimid
Formel:
C₁₄H₁₆BrNO₂ Siedepunkt: 186…188 °C/0,4mmHg Schmelzpunkt: 54…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023098 CAS-Nummer: 24566-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...