2,6-Dichlornicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichlor-3-pyridincarbonsäure, 2,6-Dichlorpyridin-3-carbonsäure
OR1907-100G
OR1907-25G
OR1907-5G
APOSOR1907-100GEA
108
EUR
APOSOR1907-100G
APOSOR1907-25G
APOSOR1907-5G
2,6-Dichlornicotinsäure 97%
2,6-Dichlornicotinsäure
Formel:
C₆H₃Cl₂NO₂ Schmelzpunkt: 140…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075583 CAS-Nummer: 38496-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...