3,5-Dichlorpicolinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloropyridine-2-carboxylic acid

OR1908-1G
APOSOR1908-1GEA 21.9 EUR
APOSOR1908-1G
3,5-Dichlorpicolinsäure
3,5-Dichlorpicolinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192,0000 g/mol
Schmelzpunkt: 156 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03230005
CAS-Nummer: 81719-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR