5,6-Dichlornicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,6-Dichlorpyridin-3-carbonsäure, 5,6-Dichlor-3-pyridincarbonsäure
OR1909-1G
APOSOR1909-1GEA
21.9
EUR
APOSOR1909-1G
5,6-Dichlornicotinsäure 97%
5,6-Dichlornicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Schmelzpunkt: 163…168 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075181 CAS-Nummer: 41667-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...