5-Aminonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid

OR1915-100G OR1915-25G OR1915-5G
APOSOR1915-100GEA 115 EUR
APOSOR1915-100G APOSOR1915-25G APOSOR1915-5G
5-Aminonicotinsäure
5-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138.13 g/mol
Schmelzpunkt: 293 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00129116
CAS-Nummer: 24242-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR