4-Ethoxy-3-methylbenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethoxy-3-methylphenylboronic acid

OR1925-5G OR1925-25G
APOSOR1925-5GEA 97.3 EUR
APOSOR1925-5G APOSOR1925-25G
4-Ethoxy-3-methylbenzeneboronic acid 97%
4-Ethoxy-3-methylbenzeneboronic acid
Formel: C₉H₁₃BO₃
Molecular Weight: 180.01 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659870
CAS-Nummer: 850568-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR