2-Amino-4-cyanobenzeneboronic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR1928-100MG
APOSOR1928-100MGEA 111 EUR
APOSOR1928-100MG
2-Amino-4-cyanobenzeneboronic acid hydrochloride 95%
2-Amino-4-cyanobenzeneboronic acid hydrochloride
Formel: C₇H₇BN₂O₂·HCl
Schmelzpunkt: 214…218 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD06659820
CAS-Nummer: 850568-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR