3,5-Dichlortoluol
Lieferant: Apollo Scientific
|
Achtung
|
OR19487-25G
OR19487-100G
APOSOR19487-25GEA
105
EUR
APOSOR19487-25G
APOSOR19487-100G
3,5-Dichlortoluol
3,5-Dichlortoluol
Formel:
C₇H₆Cl₂ Molecular Weight: 161.03 g/mol Siedepunkt: 78…79 °C/9mmHg Schmelzpunkt: 24…26 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035819 CAS-Nummer: 25186-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...