tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-(hydroxymethyl)pyridine, 2-BOC protected
OR19489-1G
APOSOR19489-1GEA
458
EUR
APOSOR19489-1G
tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate 95%
tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate
Formel:
C₁₁H₁₆N₂O₃ Schmelzpunkt: 138…139 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
877593-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...