2-Cyclopropylethylamine hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyclopropylethanamine hydrochloride

OR19507-1G
APOSOR19507-1GEA 63.8 EUR
APOSOR19507-1G
2-Cyclopropylethylamine hydrochloride 95%
2-Cyclopropylethylamine hydrochloride
Formel: C₅H₁₁N·ClH
Molecular Weight: 121,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14525749
CAS-Nummer: 89381-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR