3-Chloro-5-methylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-m-toluic acid, 3-Carboxy-5-chlorotoluene

OR19520-10G OR19520-25G
APOSOR19520-10GEA 91.5 EUR
APOSOR19520-10G APOSOR19520-25G
3-Chloro-5-methylbenzoic acid 95%
3-Chloro-5-methylbenzoic acid
Formel: C₈H₇ClO₂
Molecular Weight: 170.593 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11007823
CAS-Nummer: 56961-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR