5-Bromo-4-chloro-2-aminopyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-bromo-4-chloropyridine, 5-Bromo-4-chloropyridin-2-amine

OR19535-1G
APOSOR19535-1GEA 21.9 EUR
APOSOR19535-1G
5-Bromo-4-chloro-2-aminopyridine 97%
5-Bromo-4-chloro-2-aminopyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
Schmelzpunkt: 141…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD12407282
CAS-Nummer: 942947-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR