3-Amino-4,5-dimethylisoxazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-amino-4,5-dimethylisoxazole
OR19546-1G
APOSOR19546-1GEA
77
EUR
APOSOR19546-1G
3-Amino-4,5-dimethylisoxazole 97%
3-Amino-4,5-dimethylisoxazole
Formel:
C5H8N2O Molecular Weight: 112,1300 g/mol Schmelzpunkt: 114…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411577 CAS-Nummer: 13999-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...