(S)-Glycidyl trityl ether 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-Glycidyl Triphenylmethyl Ether, (S)-2-(Triphenylmethoxymethyl)oxirane, (S)-2-((Trityloxy)methyl)oxirane

OR19556-100G
APOSOR19556-100GEA 101 EUR
APOSOR19556-100G
(S)-Glycidyl trityl ether 98%
(S)-Glycidyl trityl ether
Formel: C₂₂H₂₀O₂
Molecular Weight: 316.40 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00273373
CAS-Nummer: 129940-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR