5,6-Dimethyl-1-indanone 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,6-Dimethyl-2,3-dihydro-1H-inden-1-one

OR19567-1G
APOSOR19567-1GEA 59.6 EUR
APOSOR19567-1G
5,6-Dimethyl-1-indanone 96%
5,6-Dimethyl-1-indanone
Formel: C₁₁H₁₂O
Molecular Weight: 160,212 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00666126
CAS-Nummer: 16440-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR