3-Amino-4-phenyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Phenyl-1H-pyrazol-3-amine

OR1969-5G
APOSOR1969-5GEA 89.9 EUR
APOSOR1969-5G
3-Amino-4-phenyl-1H-pyrazole
3-Amino-4-phenyl-1H-pyrazole
Formel: C₉H₉N₃
Molecular Weight: 159.19 g/mol
Schmelzpunkt: 174…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01693715
CAS-Nummer: 5591-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR