3-Amino-2-(4-methylphenylamino)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-N2-(4-methylphenyl)benzene-1,2-diamine

OR1980-100MG
APOSOR1980-100MGEA 65.4 EUR
APOSOR1980-100MG
3-Amino-2-(4-methylphenylamino)benzoic acid
3-Amino-2-(4-methylphenylamino)benzoic acid
Formel: C₁₄H₁₄N₂O₂
Molecular Weight: 242,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056210
CAS-Nummer: 116702-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR