3-Amino-2-(phenylamino)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR1982-100MG
APOSOR1982-100MGEA 56.7 EUR
APOSOR1982-100MG
3-Amino-2-(phenylamino)benzoic acid
3-Amino-2-(phenylamino)benzoic acid
Formel: C₁₃H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056212
CAS-Nummer: 116702-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR