tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate

Lieferant: Apollo Scientific

OR200001-1G
APOSOR200001-1GEA 0 EUR
APOSOR200001-1G
tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate
tert-Butyl-3-benzyl-3-formylpiperidine-1-carboxylate
Formel: C₁₈H₂₅NO₃
Molecular Weight: 303,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18064987
CAS-Nummer: 1355334-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR