Imidazo[1,2-a]pyridine-7-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR200003-1G
OR200003-250MG
OR200003-500MG
OR200003-2.5G
OR200003-5G
APOSOR200003-1GEA
458
EUR
APOSOR200003-1G
APOSOR200003-250MG
APOSOR200003-500MG
APOSOR200003-2.5G
APOSOR200003-5G
Imidazo[1,2-a]pyridine-7-carbaldehyde
Imidazo[1,2-a]pyridine-7-carbaldehyde
Formel:
C₈H₆N₂O Schmelzpunkt: 143…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09910326 CAS-Nummer: 136117-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...