Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Lieferant: Apollo Scientific
OR200007-1G
APOSOR200007-1GEA
0
EUR
APOSOR200007-1G
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Formel:
C₁₇H₁₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13790489 CAS-Nummer: 1157652-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...