tert-Butyl-4-[3-(methoxycarbonyl)benzyl]piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR200008-1G
APOSOR200008-1GEA 0 EUR
APOSOR200008-1G
tert-Butyl-4-[3-(methoxycarbonyl)benzyl]piperazine-1-carboxylate
tert-Butyl-4-[3-(methoxycarbonyl)benzyl]piperazine-1-carboxylate
Formel: C₁₈H₂₆N₂O₄
Molecular Weight: 334,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06808796
CAS-Nummer: 203047-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR