Ethyl-7-bromo-2-methylimidazo[1,2-a]pyridine-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR200013-1G
APOSOR200013-1GEA 406 EUR
APOSOR200013-1G
Ethyl-7-bromo-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl-7-bromo-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₁H₁₁BrN₂O₂
Molecular Weight: 283.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037416
CAS-Nummer: 81438-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR