1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl-1-(tert-butoxycarbonyl)-3-phenylpiperidine-3-carboxylate (as fumarate salt), Ethyl 3-phenylpiperidine-3-carboxylate, N-BOC protected (2E)-but-2-ene-1,4-dioate, 1-(tert-Butoxycarbonyl)-3-(ethoxycarbonyl)-3-phenylpiperidine (2E)-but-2-ene-1,4-dioate

OR200018-1G
APOSOR200018-1GEA 0 EUR
APOSOR200018-1G
1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate 90%
1-tert-Butyl 3-ethyl 3-phenylpiperidine-1,3-dicarboxylate fumarate
Formel: C₁₉H₂₇NO₄·C₄H₄O₄
Molecular Weight: 449,494 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21333141
CAS-Nummer: 1980007-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR