4-Nitro-1,3-benzoxazol-2(3H)-one
Lieferant: Apollo Scientific
Synonyme:
4-Nitrobenzo[d]oxazol-2(3H)-one
OR200021-1G
APOSOR200021-1GEA
140
EUR
APOSOR200021-1G
4-Nitro-1,3-benzoxazol-2(3H)-one
4-Nitro-1,3-benzoxazol-2(3H)-one
Formel:
C₇H₄N₂O₄ Molecular Weight: 180,12 g/mol Schmelzpunkt: 227…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00460557 CAS-Nummer: 28955-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...