1,1'-Biphenyl-2,2'-diamine

Lieferant: Apollo Scientific

Synonyme: 2,2'-Diaminobiphenyl, [1,1'-Biphenyl]-2,2'-diamine

OR200022-1G OR200022-250MG OR200022-500MG OR200022-2.5G
APOSOR200022-1GEA 68.3 EUR
APOSOR200022-1G APOSOR200022-250MG APOSOR200022-500MG APOSOR200022-2.5G
1,1'-Biphenyl-2,2'-diamine
1,1'-Biphenyl-2,2'-diamine
Formel: C₁₂H₁₂N₂
Molecular Weight: 184.24 g/mol
Schmelzpunkt: 81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00034060
CAS-Nummer: 1454-80-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR