6,7-Dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: Salsolidine HCl

OR200031-1G
APOSOR200031-1GEA 179 EUR
APOSOR200031-1G
6,7-Dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
6,7-Dimethoxy-1-methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formel: C₁₂H₁₈ClNO₂
Molecular Weight: 243,74 g/mol
Schmelzpunkt: 189…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00050647
CAS-Nummer: 63283-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR