N-Acetyl-DL-2-phenylglycine
Lieferant: Apollo Scientific
OR200032-1G
APOSOR200032-1GEA
0
EUR
APOSOR200032-1G
N-Acetyl-DL-2-phenylglycine
N-Acetyl-DL-2-phenylglycine
Formel:
C₁₀H₁₁NO₃ Schmelzpunkt: 179…199 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055924 CAS-Nummer: 15962-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...