Methyl-3-(pyrrolidin-1-ylmethyl)benzoate

Lieferant: Apollo Scientific
Achtung

OR200036-1G
APOSOR200036-1GEA 0 EUR
APOSOR200036-1G
Methyl-3-(pyrrolidin-1-ylmethyl)benzoate
Methyl-3-(pyrrolidin-1-ylmethyl)benzoate
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909459
CAS-Nummer: 321198-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR