N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine

Lieferant: Apollo Scientific

OR200039-1G
APOSOR200039-1GEA 458 EUR
APOSOR200039-1G
N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine
N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine
Formel: C₁₂H₁₆N₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22126063
CAS-Nummer: 925216-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR