Di-tert-butyl-2-chloropyridin-3-ylimidodicarbonate

Lieferant: Apollo Scientific

OR200040-1G
APOSOR200040-1GEA 0 EUR
APOSOR200040-1G
Di-tert-butyl-2-chloropyridin-3-ylimidodicarbonate
Di-tert-butyl-2-chloropyridin-3-ylimidodicarbonate
Formel: C₁₅H₂₁ClN₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD21496441
CAS-Nummer: 749265-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR