4-(2,4-Dinitrophenoxy)benzaldehyde

Lieferant: Apollo Scientific

OR200041-1G
APOSOR200041-1GEA 0 EUR
APOSOR200041-1G
4-(2,4-Dinitrophenoxy)benzaldehyde
4-(2,4-Dinitrophenoxy)benzaldehyde
Formel: C₁₃H₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00579130
CAS-Nummer: 794-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR