4-[(3-Nitro-2-pyridinyl)oxy]benzaldehyde

Lieferant: Apollo Scientific

OR200043-1G
APOSOR200043-1GEA 0 EUR
APOSOR200043-1G
4-[(3-Nitro-2-pyridinyl)oxy]benzaldehyde
4-[(3-Nitro-2-pyridinyl)oxy]benzaldehyde
Formel: C₁₂H₈N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD22126064
CAS-Nummer: 1158693-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR