3-(4-Nitrophenoxy)benzaldehyde

Lieferant: Apollo Scientific

OR200044-1G
APOSOR200044-1GEA 0 EUR
APOSOR200044-1G
3-(4-Nitrophenoxy)benzaldehyde
3-(4-Nitrophenoxy)benzaldehyde
Formel: C₁₃H₉NO₄
Molecular Weight: 243.22 g/mol
Schmelzpunkt: 84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18207479
CAS-Nummer: 17076-72-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR