3-(4-Nitrophenoxy)benzoic acid
Lieferant: Apollo Scientific
OR200045-1G
APOSOR200045-1GEA
519
EUR
APOSOR200045-1G
3-(4-Nitrophenoxy)benzoic acid
3-(4-Nitrophenoxy)benzoic acid
Formel:
C₁₃H₉NO₅ Schmelzpunkt: 188 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00690134 CAS-Nummer: 27237-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...