Ethyl-1-(2-chloropyrimidin-4-yl)piperidine-3-carboxylate

Lieferant: Apollo Scientific

OR200046-1G
APOSOR200046-1GEA 0 EUR
APOSOR200046-1G
Ethyl-1-(2-chloropyrimidin-4-yl)piperidine-3-carboxylate
Ethyl-1-(2-chloropyrimidin-4-yl)piperidine-3-carboxylate
Formel: C₁₂H₁₆ClN₃O₂
Molecular Weight: 269,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22126068
CAS-Nummer: 1347757-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR