4-(2,4-Dinitrophenyl)morpholine
Lieferant: Apollo Scientific
OR200048-1G
APOSOR200048-1GEA
0
EUR
APOSOR200048-1G
4-(2,4-Dinitrophenyl)morpholine
4-(2,4-Dinitrophenyl)morpholine
Formel:
C₁₀H₁₁N₃O₅ Schmelzpunkt: 117…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00193779 CAS-Nummer: 39242-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...