N-Benzyl-N-(2,4-dinitrophenyl)amine

Lieferant: Apollo Scientific

Synonyme: N-Benzyl-2,4-dinitroaniline

OR200049-1G
APOSOR200049-1GEA 0 EUR
APOSOR200049-1G
N-Benzyl-N-(2,4-dinitrophenyl)amine
N-Benzyl-N-(2,4-dinitrophenyl)amine
Formel: C₁₃H₁₁N₃O₄
Molecular Weight: 273,25 g/mol
Schmelzpunkt: 115…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00179575
CAS-Nummer: 7403-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR