4-(4-Nitrophenoxy)benzonitrile

Lieferant: Apollo Scientific

OR200050-1G
APOSOR200050-1GEA 0 EUR
APOSOR200050-1G
4-(4-Nitrophenoxy)benzonitrile
4-(4-Nitrophenoxy)benzonitrile
Formel: C₁₃H₈N₂O₃
Schmelzpunkt: 156…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00443712
CAS-Nummer: 17076-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR