3-(4-Nitrophenoxy)benzonitrile

Lieferant: Apollo Scientific

OR200051-1G
APOSOR200051-1GEA 0 EUR
APOSOR200051-1G
3-(4-Nitrophenoxy)benzonitrile
3-(4-Nitrophenoxy)benzonitrile
Formel: C₁₃H₈N₂O₃
Schmelzpunkt: 106…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19289635
CAS-Nummer: 17076-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR