4-Benzylmorpholine-2-carboxylic acid

Lieferant: Apollo Scientific

OR200056-1G
APOSOR200056-1GEA 0 EUR
APOSOR200056-1G
4-Benzylmorpholine-2-carboxylic acid
4-Benzylmorpholine-2-carboxylic acid
Formel: C₁₂H₁₅NO₃
Molecular Weight: 221,250 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410628
CAS-Nummer: 769087-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR