N1-Methyl-4-nitro-1,2-phenylenediamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Diamino-N1-methyl-4-nitrobenzene, N1-Methyl-4-nitrobenzene-1,2-diamine, 2-Amino-1-methylamino-4-nitrobenzene
OR200062-5G
APOSOR200062-5GEA
29
EUR
APOSOR200062-5G
N1-Methyl-4-nitro-1,2-phenylenediamine
N1-Methyl-4-nitro-1,2-phenylenediamine
Formel:
C₇H₉N₃O₂ Molecular Weight: 167,17 g/mol Schmelzpunkt: 168…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156607 CAS-Nummer: 41939-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...