2-Ethylbenzoylchlorid

Lieferant: Apollo Scientific

OR200067-1G
APOSOR200067-1GEA 0 EUR
APOSOR200067-1G
2-Ethylbenzoylchlorid
2-Ethylbenzoylchlorid
Formel: C₉H₉ClO
Molecular Weight: 168.62 g/mol
Siedepunkt: 98…100 °C/15mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 76118-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR