1-(Methylsulphonyl)piperidin-4-ol

Lieferant: Apollo Scientific
Achtung

OR200068-1G
APOSOR200068-1GEA 0 EUR
APOSOR200068-1G
1-(Methylsulphonyl)piperidin-4-ol
1-(Methylsulphonyl)piperidin-4-ol
Formel: C₆H₁₃NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD11044218
CAS-Nummer: 141482-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR