2-Amino-4-phenylthiazol 97%

Lieferant: Apollo Scientific

Synonyme: 4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine

OR200076-1G
APOSOR200076-1GEA 43.5 EUR
APOSOR200076-1G
2-Amino-4-phenylthiazol 97%
2-Amino-4-phenylthiazole
Formel: C₉H₈N₂S
Molecular Weight: 176,24 g/mol
Schmelzpunkt: 148…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039680
CAS-Nummer: 2010-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR