N-Benzyl-N-(4-nitrophenyl)amine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N-Benzyl-4-nitroaniline
OR200077-1G
APOSOR200077-1GEA
0
EUR
APOSOR200077-1G
N-Benzyl-N-(4-nitrophenyl)amine
N-Benzyl-N-(4-nitrophenyl)amine
Formel:
C₁₃H₁₂N₂O₂ Molecular Weight: 228.25 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126504 CAS-Nummer: 14309-92-3 EINECS: 238-249-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...