1-Methoxy-4-(4-nitrophenoxy)benzene

Lieferant: Apollo Scientific

OR200078-1G
APOSOR200078-1GEA 0 EUR
APOSOR200078-1G
1-Methoxy-4-(4-nitrophenoxy)benzene
1-Methoxy-4-(4-nitrophenoxy)benzene
Formel: C₁₃H₁₁NO₄
Schmelzpunkt: 109…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00443498
CAS-Nummer: 6337-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR