(2E)-3-(4-Ethoxyphenyl)acryloyl chloride

Lieferant: Apollo Scientific
Gefahr

OR200085-1G
APOSOR200085-1GEA 0 EUR
APOSOR200085-1G
(2E)-3-(4-Ethoxyphenyl)acryloyl chloride
(2E)-3-(4-Ethoxyphenyl)acryloyl chloride
Formel: C₁₁H₁₁ClO₂
Molecular Weight: 210,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11108003
CAS-Nummer: 77251-75-3
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR